Distillation therm., similar to DIN 12779, +100+250:1°C, enclosed scale, cap. prismatic, colourless, gallium filled, total immersion , length 210mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259610-GA
Supplier: Amarell
Frequently Bought Together