Distillation therm., similar to DIN, 0+100:0,5°C, enclosed scale, cap. prismatic, colourless, red special liquid, total immersion, l ength 260mm, pin-top finish with ring 1 * 1 items
Catalog # AMARD259604-FL
Supplier: Amarell
Frequently Bought Together