Thermometer similar to AFNOR, STL/2/0/600, 0+600:2°C, solid stem, frosted back, yellow backed, gallium filled, 375mm long, total imm ersion, durable pigment, suitable for government verification 1 * 1 items
Catalog # AMARA411098-GA
Supplier: Amarell
Frequently Bought Together