ContactUsLinkComponent
Thermometer similar to AFNOR, STL/1/-30/270, -30+270:1°C, solid stem, white backed, gallium filled, 375mm long, total immersion, dur able pigment, suitable for government verification 1 * 1 items
Catalog # AMARA411088-GA
Supplier:  Amarell
Thermometer similar to AFNOR, STL/1/-30/270, -30+270:1°C, solid stem, white backed, gallium filled, 375mm long, total immersion, dur able pigment, suitable for government verification 1 * 1 items
Catalog # AMARA411088-GA
Supplier:  Amarell
Frequently Bought Together