Thermometer similar to AFNOR, STL/0,5/-30/120, -35+115:0,5°C, solid stem, white backed, capillary tube specially coated inside, wit h non-wetting blue special liquid, 375mm long, total immersion, durable pigment, null
Catalog # AMARA411070-CB
Supplier: Amarell
Frequently Bought Together