ContactUsLinkComponent
Thermometer similar to BS 593, B60C/100, -20+60:0,2C, solid stem, white backed, capillary tube specially coated inside, with non-w etting blue special liquid, 400mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403254-CB
Supplier:  Amarell
Thermometer similar to BS 593, B60C/100, -20+60:0,2C, solid stem, white backed, capillary tube specially coated inside, with non-w etting blue special liquid, 400mm long, 100mm immersion, durable pigment, null
Catalog # AMARA403254-CB
Supplier:  Amarell
Frequently Bought Together