ContactUsLinkComponent
Pocket saccharimeter, 14-21:0,1%mas., with therm. 0+30:1C with red or blue special liquid, with correction scale, reading below, re f. temp. 20C, 275mm long 1 * 1 items
Catalog # AMARH847474-FL
Supplier:  Amarell
Pocket saccharimeter, 14-21:0,1%mas., with therm. 0+30:1C with red or blue special liquid, with correction scale, reading below, re f. temp. 20C, 275mm long 1 * 1 items
Catalog # AMARH847474-FL
Supplier:  Amarell
Frequently Bought Together