ContactUsLinkComponent
Hartong malt saccharimeter, 6-7:0,01% mas., with therm. +5+27:0,2°C with red or blue special liquid, with correction scale, reading below, ref. temp. 20°C, 400 mm long 1 * 1 items
Catalog # AMARH847410-FL
Supplier:  Amarell
Hartong malt saccharimeter, 6-7:0,01% mas., with therm. +5+27:0,2°C with red or blue special liquid, with correction scale, reading below, ref. temp. 20°C, 400 mm long 1 * 1 items
Catalog # AMARH847410-FL
Supplier:  Amarell
Frequently Bought Together