Thermometer similar to BS 593, A70C/total, 40+70:0,1C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 400mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA403204-CB
Supplier: Amarell
Frequently Bought Together