ContactUsLinkComponent
Thermometer similar to BS 1365, SB15C/total, 5+15:0,1°C, solid stem, white backed, capillary tube specially coated inside, with non -wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402544-CB
Supplier:  Amarell
Thermometer similar to BS 1365, SB15C/total, 5+15:0,1°C, solid stem, white backed, capillary tube specially coated inside, with non -wetting blue special liquid, 235mm long, total immersion, durable pigment, null
Catalog # AMARA402544-CB
Supplier:  Amarell
Frequently Bought Together