ContactUsLinkComponent
Thermometer similar to BS 1704, G/75, 0+360:2°C, solid stem, white backed, gallium filled, 75mm immersion, 300mm long, durable pigme nt 1 * 1 items
Catalog # AMARA402112-GA
Fournisseur:  Amarell
Thermometer similar to BS 1704, G/75, 0+360:2°C, solid stem, white backed, gallium filled, 75mm immersion, 300mm long, durable pigme nt 1 * 1 items
Catalog # AMARA402112-GA
Fournisseur:  Amarell
Frequently Bought Together