ContactUsLinkComponent
Thermometer similar to BS 1704, H/total, 0+500:5°C, solid stem, white backed, gallium filled, total immersion, 345mm long, durable p igment 1 * 1 items
Catalog # AMARA402094-GA
Supplier:  Amarell
Thermometer similar to BS 1704, H/total, 0+500:5°C, solid stem, white backed, gallium filled, total immersion, 345mm long, durable p igment 1 * 1 items
Catalog # AMARA402094-GA
Supplier:  Amarell
Frequently Bought Together