ContactUsLinkComponent
Thermometer similar to BS 1704, C/total, 0+60:0,5°C, solid stem, white backed, red special liquid, total immersion, 300mm long, dura ble pigment 1 * 1 items
Catalog # AMARA402084-FL
Supplier:  Amarell
Thermometer similar to BS 1704, C/total, 0+60:0,5°C, solid stem, white backed, red special liquid, total immersion, 300mm long, dura ble pigment 1 * 1 items
Catalog # AMARA402084-FL
Supplier:  Amarell
Frequently Bought Together