Order Entry
Belgium
Orders LinkContactUsLinkComponent
Specific Gravity Hydrometers

Specific Gravity Hydrometers

Specific gravity hydrometer, 132 H - 62, 1,350 - 1,400 °SG, 0,001 °SG, 260 mm
  AMARH826924
 :   Amarell
Specific Gravity Hydrometers
  AMARH826924
 :  Amarell
Specific Gravity Hydrometers

Specific gravity hydrometer, 132 H - 62, 1,350 - 1,400 °SG, 0,001 °SG, 260 mm

 
54,10 €
   
1 * 1 ST
 

These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.

  • Capillary prismatic
  • Reliable and simple
  • Paper scale with type designation
  • Shot weighted body

Delivery information: Supplied in a transparent plastic case.

 
  • Description:
    Specific gravity hydrometer
  • Length:
    260 mm
  • Seq:
    17
  • Division:
    0,001 °SG
  • Model:
    132 H - 62
  • Range:
    1,350 - 1,400 °SG
  • Cat. No.:
    AMARH826924
  • Accuracy:
    ±1 scale division
  • Reference temperature:
    60/60 °F (except model AMARH826764-FL: 15 °C)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.