Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Specific Gravity Hydrometers

Specific gravity hydrometer, 104 H - 62, 0,750 - 0,800 °SG, 0,001 °SG, 260 mm
N° de catalogue AMARH826784
Fournisseur:   Amarell
Specific Gravity Hydrometers
N° de catalogue AMARH826784
Fournisseur:  Amarell
Specific Gravity Hydrometers

Specific gravity hydrometer, 104 H - 62, 0,750 - 0,800 °SG, 0,001 °SG, 260 mm

Prix
45,70 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.

  • Capillary prismatic
  • Reliable and simple
  • Paper scale with type designation
  • Shot weighted body

 : Supplied in a transparent plastic case.

Spécifications
  • Désignation:
    Specific gravity hydrometer
  • Longueur:
    260 mm
  • Seq:
    05
  • Division:
    0,001 °SG
  • Modèle:
    104 H - 62
  • Plage de mesure:
    0,750 - 0,800 °SG
  • Cat. No.:
    AMARH826784
  • Précision:
    ±1 scale division
  • Température de référence:
    60/60 °F (except model AMARH826764-FL: 15 °C)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.