Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR® Premium Plus, Slide Boxes

VWR® Premium Plus, Slide Boxes

Slide boxes, Premium Plus, Red, PP, 100, 195×207×32 mm
Catalog # 631-1245
Supplier:   VWR Collection
VWR® Premium Plus, Slide Boxes
Catalog # 631-1245
Supplier:  VWR Collection
VWR® Premium Plus, Slide Boxes
VWR® Premium Plus, Slide Boxes

Slide boxes, Premium Plus, Red, PP, 100, 195×207×32 mm

Price
97,10 €
Log in for availability
1 * 5 ST
Product Details & Documents

These slide boxes feature multiple labelling areas, and an index card can be used as the lid is transparent.

  • Slide rail design ensures a secure fit
  • Central numbering grid for slide organisation and easy viewing
  • Plastic design eliminates potential rusting of metal parts
  • Sliding lock provides secure storage
  • Hinged lid smoothly opens 180°, sits flat on surface preventing misplacement
  • EVA foam lines the bottom of the box
Specifications
  • No. of slides:
    100
  • Colour:
    Red
  • Description:
    Slide boxes, Premium Plus
  • Material:
    PP
  • W×D×H:
    195×207×32 mm
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.