VWR® Premium Plus, Slide Boxes
Replacement inventory cards
Catalog # 631-1248
Supplier: VWR Collection
Jump to:
Product Details & Documents
These slide boxes feature multiple labelling areas, and an index card can be used as the lid is transparent.
- Slide rail design ensures a secure fit
- Central numbering grid for slide organisation and easy viewing
- Plastic design eliminates potential rusting of metal parts
- Sliding lock provides secure storage
- Hinged lid smoothly opens 180°, sits flat on surface preventing misplacement
- EVA foam lines the bottom of the box
Documents
VWR® Collection Selection Catalogue VWR® for Microscopy
Product Literature
Specifications
- Description:Replacement inventory cards
- Seq:101