Dip slides
Contact slide 9 PCA + TTC + Neutralising×2, 20
Catalog # LIOF525322
Supplier: LIOFILCHEM
Jump to:
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
- ISO 18593
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:20
- Description:Contact slide 9 PCA + TTC + Neutralising×2