Order Entry
Belgium
Orders LinkContactUsLinkComponent
BD® Whitacre Spinal Needles

BD® Whitacre Spinal Needles

Spinal needles with 20 G, 32 mm introducer, 25 G, 90 mm, Sterile, Blue
Catalog # BDAM405076
Supplier:   BD Medical
BD® Whitacre Spinal Needles
Catalog # BDAM405076
Supplier:  BD Medical
BD® Whitacre Spinal Needles

Spinal needles with 20 G, 32 mm introducer, 25 G, 90 mm, Sterile, Blue

Price
1 270,00 €
Log in for availability
1 * 200 ST
Product Details & Documents

The BD® Whitacre needles are manufactured with proprietary needle tipping, side-port and low-glare hub technologies.

  • Better post-operative patient experience with BD® Whitacre spinal needles that minimise the risk of postdural puncture headache (PDPH)
  • Enhanced precision with BD® Whitacre spinal needles that enable visualisation of CSF, aiding proper needle placement
  • BD® Whitacre spinal needles yield a distinct tactile 'click' or 'pop' as the pencil-point penetrates the dura, alerting the clinician of proper placement
  • Precision-formed side hole helps directional flow of anaesthetic agents and helps reduce the possibility of straddling the dura
  • The BD® Whitacre Spinal Needle is supported by 25+ independant published studies
Specifications
  • Colour code:
    Blue
  • Gauge:
    25 G
  • Type:
    Sterile
  • Description:
    Spinal needles with 20 G, 32 mm introducer
  • Length:
    90 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.