BD® Whitacre Spinal Needles
Spinal needle, 22 G, 90 mm, Sterile, Black
Catalog # BDAM401995
Supplier: BD Medical
Jump to:
Product Details & Documents
The BD® Whitacre needles are manufactured with proprietary needle tipping, side-port and low-glare hub technologies.
- Better post-operative patient experience with BD® Whitacre spinal needles that minimise the risk of postdural puncture headache (PDPH)
- Enhanced precision with BD® Whitacre spinal needles that enable visualisation of CSF, aiding proper needle placement
- BD® Whitacre spinal needles yield a distinct tactile 'click' or 'pop' as the pencil-point penetrates the dura, alerting the clinician of proper placement
- Precision-formed side hole helps directional flow of anaesthetic agents and helps reduce the possibility of straddling the dura
- The BD® Whitacre Spinal Needle is supported by 25+ independant published studies
Specifications
- Colour code:Black
- Gauge:22 G
- Type:Sterile
- Description:Spinal needle
- Length:90 mm
- Seq:02