NALG2007-0064 2000 mL rectangular bottle only, HDPE (63 mm PP cap removed and used to produce by LP/AC). Materials Supplied By: EP. Process: Particulate Cleaning & Gamma Irradiation (25-40 kGy). Packaging: Single cleanroom bag each bottle. 12 per cas 1 * 12 items
Catalog # NALGCENLV7L2PI-E
Supplier: Thermo Fisher Scientific
Frequently Bought Together