Low temperature covers
Low temperature cover, plexiglass, 6 blocks, 318×216×41 mm
Catalog # OHAU30400181
Supplier: OHAUS
Jump to:
Product Details & Documents
Plexiglass cover reduces air flow for additional temperature stability in low temperature applications.
- Two sides are open 6,4 mm
Specifications
- Description:Low temperature cover, plexiglass
- W×D×H:318×216×41 mm
- For:6 blocks



