Low temperature covers
Low temperature cover, plexiglass, 2 blocks, 165×165×41 mm
Catalog # OHAU30400179
Supplier: OHAUS
Jump to:
Product Details & Documents
Plexiglass cover reduces air flow for additional temperature stability in low temperature applications.
- Two sides are open 6,4 mm
Specifications
- Description:Low temperature cover, plexiglass
- W×D×H:165×165×41 mm
- For:2 blocks
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



