Modular heating blocks
Single block, Ø 17,1 mm, 15 ml test tubes, 12, 44,5 mm, 95×76×51 mm
Catalog # OHAU30400172
Supplier: OHAUS
Jump to:
Product Details & Documents
Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Caution: To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.
Specifications
- No. of Holes:12
- Description:Single block
- Seq:039
- W×D×H:95×76×51 mm
- Depth:44,5 mm
- Well size:Ø 17,1 mm
- For:15 ml test tubes
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



