Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Modular heating blocks

Single block, Ø 35,0 mm, 35 mm test tubes, 4, 47,6 mm, 95×76×51 mm
Catalog # OHAU30400167
Supplier:   OHAUS
Modular heating blocks
Catalog # OHAU30400167
Supplier:  OHAUS
Modular heating blocks
Modular heating blocks

Single block, Ø 35,0 mm, 35 mm test tubes, 4, 47,6 mm, 95×76×51 mm

Price
149,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.

 : To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.

Specifications
  • No. of Holes:
    4
  • Description:
    Single block
  • Seq:
    011
  • W×D×H:
    95×76×51 mm
  • Depth:
    47,6 mm
  • Well size:
    Ø 35,0 mm
  • For:
    35 mm test tubes
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.