Modular heating blocks
Single block, Ø 11,5 mm, 6 mm test tubes, 30, — mm, 95×76×51 mm
Catalog # OHAU30400163
Supplier: OHAUS
Jump to:
Product Details & Documents
Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Caution: To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.
Specifications
- No. of Holes:30
- Description:Single block
- Seq:033
- W×D×H:95×76×51 mm
- Depth:— mm
- Well size:Ø 11,5 mm
- For:6 mm test tubes
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



