Order Entry
Belgium
Orders LinkContactUsLinkComponent
Modular heating blocks

Modular heating blocks

Single block, Ø 35,0 mm, 1,5 ml Eppendorf microtubes, 20, 36,9 mm, 95×76×51 mm
Catalog # OHAU30400162
Supplier:   OHAUS
Modular heating blocks
Catalog # OHAU30400162
Supplier:  OHAUS
Modular heating blocks
Modular heating blocks

Single block, Ø 35,0 mm, 1,5 ml Eppendorf microtubes, 20, 36,9 mm, 95×76×51 mm

Price
149,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.

Caution: To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.

Specifications
  • No. of Holes:
    20
  • Description:
    Single block
  • Seq:
    035
  • W×D×H:
    95×76×51 mm
  • Depth:
    36,9 mm
  • Well size:
    Ø 35,0 mm
  • For:
    1,5 ml Eppendorf microtubes