Modular heating blocks
Single block, Ø 7,9; 11,1; 11,5 mm, 6×0,5, 21, 27,6; 39,1; 38,1 mm, 95×76×51 mm
Catalog # OHAU30400194
Supplier: OHAUS
Jump to:
Product Details & Documents
Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Caution: To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.
Specifications
- No. of Holes:21
- Description:Single block
- W×D×H:95×76×51 mm
- Depth:27,6; 39,1; 38,1 mm
- Well size:Ø 7,9; 11,1; 11,5 mm
- For:6×0,5



