Modular heating blocks
Single block, Ø 11,1; 17,1; 29,0 mm, 1,5, 9, 39,1; 44,5; 47,6 mm, 95×76×51 mm
Catalog # OHAU30400193
Supplier: OHAUS
Jump to:
Product Details & Documents
Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Caution: To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.
Specifications
- No. of Holes:9
- Description:Single block
- Seq:020
- W×D×H:95×76×51 mm
- Depth:39,1; 44,5; 47,6 mm
- Well size:Ø 11,1; 17,1; 29,0 mm
- For:1,5
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



