Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Modular heating blocks

Single block, Ø 11,5 mm, 2 ml Eppendorf microtubes, 20, 38,1 mm, 95×76×51 mm
Catalog # OHAU30400191
Supplier:   OHAUS
Modular heating blocks
Catalog # OHAU30400191
Supplier:  OHAUS
Modular heating blocks
Modular heating blocks

Single block, Ø 11,5 mm, 2 ml Eppendorf microtubes, 20, 38,1 mm, 95×76×51 mm

Price
149,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.

 : To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.

Specifications
  • No. of Holes:
    20
  • Description:
    Single block
  • Seq:
    034
  • W×D×H:
    95×76×51 mm
  • Depth:
    38,1 mm
  • Well size:
    Ø 11,5 mm
  • For:
    2 ml Eppendorf microtubes