pH Buffer Solution pH 9.21 ± 0.02 ,25°C (Coloured) 1 * 1 L
Catalog # REGETB9211
Supplier: REAGECON
Frequently Bought Together
VWR®, Glass Fibre Filters
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Redox (ORP) Standards
Supplier: VWR Chemicals
Trisodium citrate, anhydrous 99% (by titrimetric analysis)
Supplier: Thermo Fisher Scientific