Order Entry
Belgium
Orders LinkContactUsLinkComponent
ω-Bromo-2-nitroacetophenone

ω-Bromo-2-nitroacetophenone

Bottle, 5 g
Catalog # APOSOR2385-5G
Corrosive
Supplier:   Apollo Scientific
ω-Bromo-2-nitroacetophenone
Catalog # APOSOR2385-5G
Supplier:  Apollo Scientific
CAS Number:  6851-99-6
ω-Bromo-2-nitroacetophenone

Bottle, 5 g

CAS Number:  6851-99-6
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  α-Bromo-2-nitroacetophenone, 2-Bromo-2'-nitroacetophenone, Bromomethyl o-nitrophenyl ketone, 2-Nitrophenacyl bromide
  • CAS Number:6851-99-6
  • Formula:C₈H₆BrNO₃
  • MDL Number:MFCD00010294
  • Molecular Weight:244.04 g/mol
  • Melting Pt:53…57 °C
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.