Order Entry
Belgium
Orders LinkContactUsLinkComponent
ω-Bromo-2-nitroacetophenone

ω-Bromo-2-nitroacetophenone

Bottle, 1 g
Catalog # APOSOR2385-1G
Corrosif
Supplier:   Apollo Scientific
ω-Bromo-2-nitroacetophenone
Catalog # APOSOR2385-1G
Supplier:  Apollo Scientific
CAS Number:  6851-99-6
ω-Bromo-2-nitroacetophenone

Bottle, 1 g

CAS Number:  6851-99-6
Price
21,80 €
Connexion Pour la disponibilité
1 * 1 g
Product Details & Documents
Synonymes:  α-Bromo-2-nitroacetophenone, 2-Bromo-2'-nitroacetophenone, Bromomethyl o-nitrophenyl ketone, 2-Nitrophenacyl bromide
  • Numéro CAS:6851-99-6
  • Formule:C₈H₆BrNO₃
  • Numéro MDL:MFCD00010294
  • Poids moléculaire:244.04 g/mol
  • Point de fusion:53…57 °C
  • Température de stockage:Ambient
  • UN:3261
  • ADR:8,II
Spécifications

Spécifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.