Order Entry
Belgium
Orders LinkContactUsLinkComponent
Curcumin 97%

Curcumin 97%

Bottle, 100 g
Catalog # APOSOR24598-100G
Irritant
Supplier:   Apollo Scientific
Curcumin 97%
Catalog # APOSOR24598-100G
Supplier:  Apollo Scientific
CAS Number:  458-37-7
Curcumin 97%

Bottle, 100 g

CAS Number:  458-37-7
Price
134,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
  • CAS Number:458-37-7
  • Formula:C₂₁H₂₀O₆
  • MDL Number:MFCD00008365
  • Molecular Weight:368.39 g/mol
  • Melting Pt:170…175 °C
  • Storage Temperature:Freezer
  • EINECS:207-280-5

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.