Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Fluorophenol 99%

4-Fluorophenol 99%

Bottle, 500 g
Catalog # APOSPC4090-500G
Corrosive
Toxic
Supplier:   Apollo Scientific
4-Fluorophenol 99%
Catalog # APOSPC4090-500G
Supplier:  Apollo Scientific
CAS Number:  371-41-5
4-Fluorophenol 99%

Bottle, 500 g

CAS Number:  371-41-5
Price
68,10 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185 °C
  • Melting Pt:44…47 °C
  • Density:1.22 g/cm³ (25 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:206-736-0
  • UN:2928
  • ADR:6.1+8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.