Valinomycin
Bottle, 100 mg
Catalog # APOSBIV0893-100MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Anitbiotic produced by Streptomyces fulvissimus. Valinomycin is highly selective for K+ ions over Na+without influencing the permeability of other ions
- CAS Number:2001-95-8
- Formula:C₅₄H₉₀O₁₈N₆
- MDL Number:MFCD00005114
- Molecular Weight:1111.34 g/mol
- Melting Pt:187…190 °C
- Storage Temperature:Refrigerator
- EINECS:217-896-6
- UN:2811
- ADR:6.1,I
Specifications
Specifications
- Pack type:Bottle
- Pk:100 mg



