Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)

Tween® 20 (Polysorbate)

Bottle, 1 L
Catalog # APOSBIT0803-1L
Supplier:   Apollo Scientific
Tween® 20 (Polysorbate)
Catalog # APOSBIT0803-1L
Supplier:  Apollo Scientific
CAS Number:  9005-64-5
Tween® 20 (Polysorbate)

Bottle, 1 L

CAS Number:  9005-64-5
Price
79,60 €
Log in for availability
1 * 1 L
Product Details & Documents

Non ionic detergent used for cell lysis, nuclei isolationand cell fractionation.

Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Boiling Pt:>100 °C
  • Melting Pt:–14 °C
  • Density:1.095 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.