Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Bottle, 1 L
Catalog # APOSBIT0501-1L
Irritant
Supplier:   Apollo Scientific
Tween® 80 (Polysorbate)
Catalog # APOSBIT0501-1L
Supplier:  Apollo Scientific
CAS Number:  9005-65-6
Tween® 80 (Polysorbate)

Bottle, 1 L

CAS Number:  9005-65-6
Price
55,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Non ionic detergent used for cell lysis, nuclei isolationand cell fractionationBulk pricing available

  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Boiling Pt:100 °C
  • Melting Pt:–25 °C
  • Density:1.064 g/cm³ (25 °C)
  • Flash Pt:>113 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.