Order Entry
Belgium
Orders LinkContactUsLinkComponent
α-Cyclodextrin

α-Cyclodextrin

Bottle, 250 g
Catalog # APOSBIM1014-250G
Supplier:   Apollo Scientific
α-Cyclodextrin
Catalog # APOSBIM1014-250G
Supplier:  Apollo Scientific
CAS Number:  10016-20-3
α-Cyclodextrin

Bottle, 250 g

CAS Number:  10016-20-3
Price
319,00 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.