Order Entry
Belgium
Orders LinkContactUsLinkComponent
Agar

Agar

Bottle, 1 kg
Catalog # APOSBIM1002-1KG
Irritant
Supplier:   Apollo Scientific
Agar
Catalog # APOSBIM1002-1KG
Supplier:  Apollo Scientific
CAS Number:  9002-18-0
Agar

Bottle, 1 kg

CAS Number:  9002-18-0
Price
123,00 €
Log in for availability
1 * 1 kg
Product Details & Documents

A purified agar with a high gel strength and clarity for tissue culture and microbiological work. Minimal conc. of 5.0g/L to obtain solid gel.

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications