Order Entry
Belgium
Orders LinkContactUsLinkComponent
Nile red

Nile red

Bottle, 1 g
Catalog # APOSBIN0465-1G
Supplier:   Apollo Scientific
Nile red
Catalog # APOSBIN0465-1G
Supplier:  Apollo Scientific
CAS Number:  7385-67-3
Nile red

Bottle, 1 g

CAS Number:  7385-67-3
Price
145,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
  • CAS Number:7385-67-3
  • Formula:C₂₀H₁₈N₂O₂
  • MDL Number:MFCD00011639
  • Molecular Weight:318.38 g/mol
  • Melting Pt:203…205 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.