Erythromycin
Bottle, 25 g
Catalog # APOSBIE0122-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Erythromycin is a macrolide antibiotic with a bacteriostatic action against primarily gram-positive bacteria.
- 960mcg/mg
Synonyms: (-)-Erythromycin
- CAS Number:114-07-8
- Formula:C₃₇H₆₇NO₁₃
- MDL Number:MFCD00084654
- Molecular Weight:733.95 g/mol
- Melting Pt:133 °C
- Storage Temperature:Ambient
- EINECS:204-040-1
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g



