Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Erythromycin

Bottle, 10 g
Catalog # APOSBIE0122-10G
Irritant
Supplier:   Apollo Scientific
Erythromycin
Catalog # APOSBIE0122-10G
Supplier:  Apollo Scientific
CAS Number:  114-07-8
Erythromycin

Bottle, 10 g

CAS Number:  114-07-8
Price
65,20 €
Log in for availability
1 * 10 g
Product Details & Documents

Erythromycin is a macrolide antibiotic with a bacteriostatic action against primarily gram-positive bacteria.

  • 960mcg/mg
Synonyms:  (-)-Erythromycin
  • CAS Number:114-07-8
  • Formula:C₃₇H₆₇NO₁₃
  • MDL Number:MFCD00084654
  • Molecular Weight:733.95 g/mol
  • Melting Pt:133 °C
  • Storage Temperature:Ambient
  • EINECS:204-040-1
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.