Order Entry
Belgium
Orders LinkContactUsLinkComponent
Imidazole

Imidazole

Bottle, 2,5 kg
Catalog # APOSBID0565-2.5KG
Corrosive
Irritant
Health
Supplier:   Apollo Scientific
Imidazole
Catalog # APOSBID0565-2.5KG
Supplier:  Apollo Scientific
CAS Number:  288-32-4
Imidazole

Bottle, 2,5 kg

CAS Number:  288-32-4
Price
78,20 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Biological buffer - useful range 6.2-7.8

  • Assay (by titration) 99%
  • White crystals
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Boiling Pt:235 °C
  • Melting Pt:85…91 °C
  • Density:1.03 g/cm³ (25 °C)
  • Flash Pt:145 °C
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:3263
  • ADR:8,II

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.