Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(-)-Aspartic acid

Bottle, 250 g
Catalog # APOSBIA0866-250G
Irritant
Supplier:   Apollo Scientific
D-(-)-Aspartic acid
Catalog # APOSBIA0866-250G
Supplier:  Apollo Scientific
CAS Number:  1783-96-6
D-(-)-Aspartic acid

Bottle, 250 g

CAS Number:  1783-96-6
Price
33,30 €
Log in for availability
1 * 250 g
Product Details & Documents

Chiral intermediate for pharmaceuticals and agrochemicals; Materials for peptides.

Synonyms:  D-Aminosuccinic acid, D(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-2-Amino-1,4-butanedioic acid, (R)-(-)-Aspartic acid, (R)-()-Aminosuccinic acid, (R)-Aminosuccinic acid, (R)-2-Aminosuccinic acid, D-Asparaginic acid, D-Aspartic acid, H-D-Asp-OH
  • CAS Number:1783-96-6
  • Formula:C₄H₇NO₄
  • MDL Number:MFCD00063081
  • Molecular Weight:133.1 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Density:1.66 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:217-234-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications