Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bromophenol blue

Bromophenol blue

Bottle, 25 g
Catalog # APOSBIB1005-25G
Irritant
Supplier:   Apollo Scientific
Bromophenol blue
Catalog # APOSBIB1005-25G
Supplier:  Apollo Scientific
CAS Number:  115-39-9
Bromophenol blue

Bottle, 25 g

CAS Number:  115-39-9
Price
86,90 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Melting Pt:279 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications