D(+)-Arabitol
Bottle, 25 g
Catalog # APOSBIA4016-25G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Producer of xylitol via biotransformation.
Synonyms: D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
- CAS Number:488-82-4
- Formula:C₅H₁₂O₅
- MDL Number:MFCD00004709
- Molecular Weight:152.15 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:25 g



