Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D(+)-Arabitol

Bottle, 100 g
Catalog # APOSBIA4016-100G
Irritant
Supplier:   Apollo Scientific
D(+)-Arabitol
Catalog # APOSBIA4016-100G
Supplier:  Apollo Scientific
CAS Number:  488-82-4
D(+)-Arabitol

Bottle, 100 g

CAS Number:  488-82-4
Price
355,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Producer of xylitol via biotransformation.

Synonyms:  D-Arabitol, (2R,4R)-Pentane-1,2,3,4,5-pentaol
  • CAS Number:488-82-4
  • Formula:C₅H₁₂O₅
  • MDL Number:MFCD00004709
  • Molecular Weight:152.15 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications