Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pipe blind lockouts

Pipe blind lockouts

Pipe blind lockout, large (hand wheel diameter 355 - 1219 mm), Red, 628,65×247,65×92,08 mm
Catalog # BRDY149230
Supplier:   Brady
Pipe blind lockouts
Catalog # BRDY149230
Supplier:  Brady
Pipe blind lockouts
Pipe blind lockouts

Pipe blind lockout, large (hand wheel diameter 355 - 1219 mm), Red, 628,65×247,65×92,08 mm

Price
154,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These devices can be applied to the widest range of pipes to easily and temporarily avoid the release of gas, steam and hazardous liquids while pipe maintenance is still ongoing.

  • Adjustable locking arm accommodates both standard and recessed bolt positions
  • Secure 1 or 2 bolts for extra security
  • Heavy duty construction withstands harsh environments
  • UL approved for use in explosive atmospheres

Min. shackle clearance: 19,05 mm
Max. shackle diameter: 7,00 mm

Specifications
  • Type:
    Pipe blind lockout, large (hand wheel diameter 355 - 1219 mm)
  • Colour:
    Red
  • Seq:
    03
  • W×D×H:
    628,65×247,65×92,08 mm
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.