Pipe blind lockouts
Pipe blind lockout, medium (hand wheel diameter 76 - 355 mm), Red, 374,65×215,9×73,02 mm
Catalog # BRDY149229
Supplier: Brady
Aller à :
Product Details & Documents
These devices can be applied to the widest range of pipes to easily and temporarily avoid the release of gas, steam and hazardous liquids while pipe maintenance is still ongoing.
- Adjustable locking arm accommodates both standard and recessed bolt positions
- Secure 1 or 2 bolts for extra security
- Heavy duty construction withstands harsh environments
- UL approved for use in explosive atmospheres
Min. shackle clearance: 19,05 mm
Max. shackle diameter: 7,00 mm
Documents
Spécifications
- Type:Pipe blind lockout, medium (hand wheel diameter 76 - 355 mm)
- Colour:Red
- Seq:02
- W×D×H:374,65×215,9×73,02 mm



